Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025495.3 | NP_079771.1 | 8 | 845 | ||
GenBank mRNA | AK006234.1 | - | 8 | 882 | ||
AK012863.1 | - | 8 | 849 | |||
BC094280.1 | - | 8 | 898 | |||
BC147207.1 | - | 8 | 825 | 9056262 | ||
BC147208.1 | - | 8 | 825 | 9056260 |