Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160208.1 | NP_001153680.1 | 5 | 922 | ||
NM_001160209.1 | NP_001153681.1 | 4 | 1009 | |||
NM_025505.4 | NP_079781.2 | 6 | 943 | |||
GenBank mRNA | AK005489.1 | - | 4 | 743 | ||
AK006544.1 | - | 5 | 926 | |||
AK029787.1 | - | 6 | 944 | |||
AK167493.1 | - | 4 | 1016 | |||
AK168933.1 | - | 4 | 770 | |||
AK171167.1 | - | 4 | 801 | |||
BC027025.1 | - | 4 | 784 | 5374749 |