Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025513.3 | NP_079789.1 | 4 | 768 | ||
GenBank mRNA | AK006281.1 | - | 4 | 664 | ||
AK009165.1 | - | 2 | 180 | |||
BC054085.1 | - | 4 | 773 | 3371550 |