Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025516.4 | NP_079792.1 | 2, 3 | 176 | ||
GenBank mRNA | AK009350.1 | - | 2, 3 | 177 | ||
AK013942.1 | - | 2, 3 | 176 | |||
AK050285.1 | - | 2, 3 | 191 | |||
BC043720.1 | - | 2, 3 | 201 | 5375947 | ||
BC057130.1 | - | 2, 3 | 195 | 6819420 |