Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025517.3 | NP_079793.2 | 10 | 1116 | ||
GenBank mRNA | AK002543.1 | - | 10 | 1026 | ||
AK009245.1 | - | 10 | 1052 | |||
AK151108.1 | - | 10 | 1004 | |||
AK151400.1 | - | 10 | 1004 | |||
AK152129.1 | - | 10 | 1117 | |||
AK159961.1 | - | 10 | 1006 | |||
AK167830.1 | - | 10 | 993 | |||
AK190960.1 | - | 4 | 272 | |||
AK193515.1 | - | 4 | 253 | |||
AK209245.1 | - | 4 | 253 | |||
AK212568.1 | - | 4 | 206 | |||
BC016519.1 | - | 10 | 1042 | 3495638 | ||
BC042473.1 | - | 1 | 307 | |||
BC051062.1 | - | 1 | 307 | |||
BC090403.1 | - | 10 | 1087 | 5293337 |