Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025517.3 | NP_079793.2 | 3 | 347 | ||
GenBank mRNA | AK002543.1 | - | 3 | 258 | ||
AK009245.1 | - | 3 | 283 | |||
AK151108.1 | - | 3 | 235 | |||
AK151400.1 | - | 3 | 235 | |||
AK152129.1 | - | 3 | 348 | |||
AK159961.1 | - | 3 | 237 | |||
AK167830.1 | - | 3 | 224 | |||
BC016519.1 | - | 3 | 273 | 3495638 | ||
BC090403.1 | - | 3 | 318 | 5293337 |