Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dus2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301176.1 | NP_001288105.1 | 11, 12 | 1005 | ||
NM_025518.4 | NP_079794.1 | 12, 13 | 1049 | |||
GenBank mRNA | AK009391.1 | - | 12, 13 | 1021 | ||
AK077451.1 | - | 11, 12 | 977 | |||
BC058431.1 | - | 3, 4 | 320 | |||
BC116934.1 | - | 12, 13 | 967 | 40126253 | ||
BC116936.1 | - | 12, 13 | 967 | 40126255 |