Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025519.2 | NP_079795.1 | 2, 3 | 436 | ||
GenBank mRNA | AK009273.1 | - | 2, 3 | 436 | ||
AK010707.1 | - | 2, 3 | 503 | |||
AK160189.1 | - | 2, 3 | 430 | |||
BC115983.1 | - | 2, 3 | 426 | 40105850 | ||
BC117538.1 | - | 2, 3 | 426 | 40105849 |