Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002341.1 | NP_002332.1 | 4 | 818 | ||
NM_009588.1 | NP_033666.1 | 3 | 772 | |||
GenBank mRNA | L11015.1 | - | 4 | 818 | ||
U89922.1 | - | 3 | 772 |