Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025527.3 | NP_079803.2 | 2 | 180 | ||
GenBank mRNA | AK004118.1 | - | 2 | 182 | ||
AK009412.1 | - | 2 | 144 | |||
AK010760.1 | - | 2 | 179 | |||
AK037009.1 | - | 2 | 182 | |||
AK135508.1 | - | 2 | 150 | |||
AK152322.1 | - | 2 | 159 | |||
AK162107.1 | - | 2 | 150 | |||
BC051932.1 | - | 2 | 147 | 5710637 |