Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsfm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025537.3 | NP_079813.1 | 6 | 651 | ||
GenBank mRNA | AJ277891.1 | - | 5 | 488 | ||
AK009898.1 | - | 6 | 672 | |||
AK012240.1 | - | 6 | 642 | |||
AK018591.1 | - | 6 | 640 | |||
AK169164.1 | - | 6 | 642 | |||
AK171656.1 | - | 6 | 648 | |||
AK172112.1 | - | 6 | 648 | |||
AK180094.1 | - | 2 | 165 | |||
BC034286.1 | - | 6 | 630 | |||
BC051508.1 | - | 3 | 232 | |||
BC057904.1 | - | 6 | 651 | 5012380 |