Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025541.3 | NP_079817.1 | 4 | 790 | ||
GenBank mRNA | AK007804.1 | - | 4 | 646 | ||
AK010210.1 | - | 4 | 800 | |||
AK012376.1 | - | 4 | 781 | |||
AK031513.1 | - | 4 | 756 | |||
AK088618.1 | - | 4 | 733 | |||
BC027628.1 | - | 4 | 582 | 1178866 |