Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcts2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025543.2 | NP_079819.1 | 1 | 347 | ||
GenBank mRNA | AK002563.1 | - | 1 | 325 | ||
AK010255.1 | - | 1 | 348 | |||
AK028001.1 | - | 1 | 326 | |||
BC027507.1 | - | 1 | 340 | 1478068 |