Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sac3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133678.3 | NP_598439.3 | 2 | 1117 | ||
GenBank mRNA | AJ131957.2 | - | 2 | 1111 | ||
AK011298.1 | - | 2 | 1029 | |||
AK014138.1 | - | 2 | 1071 | |||
AK075930.1 | - | 2 | 1077 | |||
AK139556.1 | - | 2 | 1077 | |||
BC144812.1 | - | 2 | 1181 | 9053360 | ||
BC145789.1 | - | 2 | 1181 | 40131092 |