Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025544.2 | NP_079820.2 | 7 | 640 | ||
GenBank mRNA | AB049947.1 | - | 7 | 516 | ||
AK003126.1 | - | 7 | 561 | |||
AK008623.1 | - | 7 | 592 | |||
AK010330.1 | - | 7 | 603 | |||
AK050396.1 | - | 7 | 644 | |||
BC029193.1 | - | 7 | 559 | 4457237 | ||
BC055861.1 | - | 7 | 539 | 3587953 |