Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aptx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025444.3 | NP_001020615.1 | 7 | 888 | ||
NM_001025445.2 | NP_001020616.1 | 7 | 883 | |||
NM_025545.4 | NP_079821.3 | 7 | 1023 | |||
GenBank mRNA | AK005286.1 | - | 6 | 840 | ||
AK010516.1 | - | 7 | 870 | |||
AK017522.1 | - | 6 | 851 | |||
AK077351.1 | - | 7 | 1024 | |||
AK088928.1 | - | 6 | 573 | |||
AY040780.1 | - | 7 | 1034 | |||
AY208844.1 | - | 7 | 1035 | |||
BC021872.1 | - | 7 | 881 | 3994375 | ||
BC068309.1 | - | 6 | 796 | |||
BC071213.1 | - | 7 | 875 |