Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mterf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025547.3 | NP_079823.2 | 3, 4 | 638 | ||
GenBank mRNA | AK002299.1 | - | 3, 4 | 579 | ||
AK010525.1 | - | 3, 4 | 633 | |||
AK011934.1 | - | 3, 4 | 626 | |||
AK034645.1 | - | 3, 4 | 605 | |||
BC025173.1 | - | 3, 4 | 607 | 2655114 | ||
BC085282.1 | - | 3, 4 | 608 | 30467998 |