Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbcb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025548.3 | NP_079824.2 | 5 | 936 | ||
GenBank mRNA | AK002316.1 | - | 5 | 677 | ||
AK003655.2 | - | 5 | 682 | |||
AK010438.1 | - | 5 | 693 | |||
BC010684.1 | - | 5 | 669 | 2648112 |