Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025550.2 | NP_079826.2 | 6 | 886 | ||
GenBank mRNA | AJ252147.1 | - | 6 | 922 | ||
AK002343.1 | - | 6 | 922 | |||
AK010287.1 | - | 6 | 919 | |||
AK211149.1 | - | 3 | 164 | |||
BC006869.1 | - | 6 | 886 | 2646325 |