Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025551.3 | NP_079827.2 | 4 | 407 | ||
GenBank mRNA | AK003131.1 | - | 4 | 380 | ||
AK003322.1 | - | 4 | 380 | |||
AK008876.1 | - | 4 | 380 | |||
AK010460.1 | - | 4 | 408 | |||
AK032774.1 | - | 4 | 381 | |||
AK134765.1 | - | 4 | 380 | |||
AK153240.1 | - | 4 | 380 | |||
BC004633.1 | - | 1837 | ||||
BC086922.1 | - | 4 | 412 | 6751147 |