Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025554.2 | NP_079830.2 | 5, 6 | 590 | ||
GenBank mRNA | AK015823.1 | - | 5, 6 | 588 | ||
AK088610.1 | - | 5, 6 | 647 | |||
AK132109.1 | - | 5, 6 | 645 | |||
AK144668.1 | - | 5, 6 | 651 | |||
AK145201.1 | - | 5, 6 | 654 | |||
BC005599.1 | - | 5, 6 | 642 | |||
BC037681.1 | - | 2, 3 | 1985 | |||
BC045521.1 | - | 5, 6 | 590 | 2647754 |