Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190322.2 | NP_001177251.1 | 4 | 444 | ||
NM_001190323.2 | NP_001177252.1 | 4 | 320 | |||
NM_001190324.2 | NP_001177253.1 | 4 | 275 | |||
NM_001285804.1 | NP_001272733.1 | 4 | 475 | |||
NM_181391.4 | NP_852056.1 | 4 | 489 | |||
GenBank mRNA | AA980189.1 | - | 4 | 272 | ||
AK003308.1 | - | 4 | 321 | |||
AK007844.1 | - | 4 | 464 | |||
AK013200.1 | - | 4 | 462 | |||
AK145105.1 | - | 4 | 464 | |||
AK168647.1 | - | 4 | 419 | |||
AK169018.1 | - | 4 | 464 | |||
BC030366.1 | - | 4 | 447 | 5370748 | ||
BX527959.1 | - | 4 | 236 | |||
BY578448.1 | - | 3 | 207 | |||
CB057699.1 | - | 4 | 401 |