Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025567.2 | NP_079843.1 | 5 | 738 | ||
GenBank mRNA | AK002815.1 | - | 5 | 716 | ||
AK011288.1 | - | 5 | 739 | |||
AK151299.1 | - | 5 | 729 | |||
AK170272.1 | - | 4 | 1338 | |||
AK208195.1 | - | 1 | 68 | |||
AK215025.1 | - | 1 | 68 | |||
AK215845.1 | - | 1 | 68 | |||
BC005620.1 | - | 5 | 719 | 3711323 | ||
BC082790.1 | - | 5 | 676 |