Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025575.3 | NP_079851.2 | 2 | 296 | ||
GenBank mRNA | AK003071.1 | - | 2 | 289 | ||
AK005249.1 | - | 2 | 199 | |||
AK005510.1 | - | 2 | 233 | |||
AK007882.1 | - | 2 | 297 | |||
AK009684.1 | - | 2 | 260 | |||
AK011753.1 | - | 2 | 292 | |||
AK013220.1 | - | 2 | 271 | |||
AK017524.1 | - | 1 | 129 | |||
AK032073.1 | - | 2 | 267 | |||
AK032098.1 | - | 1 | 131 | |||
AK075770.1 | - | 2 | 261 | |||
AK139605.1 | - | 1 | 467 | |||
AK148305.1 | - | 2 | 234 | |||
BC053943.1 | - | 1 | 130 | |||
BC061084.1 | - | 1 | 170 | 6596164 |