Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anp32e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253757.1 | NP_001240686.1 | 3 | 556 | ||
NM_023210.4 | NP_075699.3 | 3 | 556 | |||
GenBank mRNA | AB037685.1 | - | 3 | 231 | ||
AK012759.1 | - | 3 | 523 | |||
AK049647.1 | - | 3 | 523 | |||
AK076049.1 | - | 3 | 383 | |||
AK088401.1 | - | 3 | 536 | |||
AK147888.1 | - | 3 | 522 | |||
AK159446.1 | - | 3 | 523 | |||
AK168380.1 | - | 3 | 505 | |||
AK172146.1 | - | 3 | 570 | |||
AK193570.1 | - | 1 | 18 | |||
AK199406.1 | - | 1 | 18 | |||
AK202251.1 | - | 1 | 19 | |||
AK204175.1 | - | 1 | 18 | |||
BC005690.1 | - | 3 | 491 | 3596597 | ||
BC080684.1 | - | 3 | 496 | 6493476 | ||
CF952181.1 | - | 3 | 227 | |||
U89345.2 | - | 3 | 517 |