Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135685.1 | NP_001129157.1 | 8 | 1217 | ||
NM_002344.5 | NP_002335.2 | 9 | 1400 | |||
NM_206961.3 | NP_996844.1 | 8 | 1217 | |||
XM_011521556.2 | XP_011519858.1 | 9 | 1400 | |||
XM_011521557.2 | XP_011519859.1 | 8 | 1232 | |||
XM_017022181.1 | XP_016877670.1 | 9 | 1415 | |||
XM_017022182.1 | XP_016877671.1 | 9 | 1415 | |||
XM_017022183.1 | XP_016877672.1 | 9 | 1415 | |||
GenBank mRNA | AK296892.1 | - | 8 | 1217 | ||
AK314006.1 | - | 9 | 1367 | |||
BC045607.1 | - | 9 | 1373 | 5270700 | ||
BC143903.1 | - | 3 | 355 | |||
D16105.1 | - | 9 | 1319 | |||
X52213.1 | - | 2 | 235 | |||
X60702.1 | - | 8 | 1209 |