Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usmg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023211.4 | NP_075700.2 | 4 | 323 | ||
GenBank mRNA | AJ290947.1 | - | 331 | |||
AK008345.1 | - | 4 | 324 | |||
AK008511.1 | - | 1153 | ||||
AK038457.1 | - | 4 | 330 | |||
AK088856.1 | - | 4 | 341 | |||
BC024355.1 | - | 4 | 404 | |||
BC087919.1 | - | 4 | 323 | 6545418 |