Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135685.1 | NP_001129157.1 | 18 | 2224 | ||
NM_002344.5 | NP_002335.2 | 20 | 2614 | |||
NM_206961.3 | NP_996844.1 | 19 | 2431 | |||
XM_011521556.2 | XP_011519858.1 | 20 | 2662 | |||
XM_011521557.2 | XP_011519859.1 | 19 | 2494 | |||
XM_017022181.1 | XP_016877670.1 | 20 | 2677 | |||
XM_017022182.1 | XP_016877671.1 | 19 | 2533 | |||
XM_017022183.1 | XP_016877672.1 | 19 | 2422 | |||
GenBank mRNA | AK296892.1 | - | 18 | 2224 | ||
AK314006.1 | - | 20 | 2581 | |||
BC045607.1 | - | 18 | 2931 | 5270700 | ||
BC143903.1 | - | 14 | 1617 | |||
CA394397.1 | - | 1 | 55 | |||
D16105.1 | - | 20 | 2533 | |||
X52213.1 | - | 13 | 1494 | |||
X60702.1 | - | 19 | 2423 |