Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppdpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025598.2 | NP_079874.1 | 4 | 449 | ||
GenBank mRNA | AK002602.1 | - | 3 | 569 | ||
AK003282.1 | - | 4 | 450 | |||
AK012037.1 | - | 4 | 333 | |||
AK012327.1 | - | 4 | 451 | |||
AK012545.1 | - | 2 | 424 | |||
AK014211.1 | - | 3 | 403 | |||
AK154758.1 | - | 4 | 490 | |||
BC029225.1 | - | 3 | 385 | |||
BC058516.1 | - | 4 | 487 | 6824261 | ||
BC091753.1 | - | 4 | 256 | 4218108 | ||
BC100439.1 | - | 4 | 341 | 6531448 |