Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppdpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025598.2 | NP_079874.1 | 5 | 527 | ||
GenBank mRNA | AK002602.1 | - | 4 | 647 | ||
AK003282.1 | - | 5 | 528 | |||
AK012037.1 | - | 5 | 411 | |||
AK012327.1 | - | 5 | 529 | |||
AK012545.1 | - | 3 | 502 | |||
AK014211.1 | - | 4 | 481 | |||
AK154758.1 | - | 5 | 568 | |||
AK166314.1 | - | 491 | ||||
BC029225.1 | - | 4 | 463 | |||
BC058516.1 | - | 5 | 565 | 6824261 | ||
BC091753.1 | - | 5 | 334 | 4218108 | ||
BC100439.1 | - | 5 | 419 | 6531448 |