Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025599.3 | NP_079875.2 | 10 | 905 | ||
GenBank mRNA | AK012168.1 | - | 10 | 886 | ||
AK019979.1 | - | 10 | 881 | |||
BC038891.1 | - | 10 | 792 | 3371329 |