Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700029H14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080781.2 | NP_001074250.1 | 1, 2 | 300 | ||
NM_025601.3 | NP_079877.1 | 1, 2 | 300 | |||
GenBank mRNA | AK006340.1 | - | 1, 2 | 245 | ||
AK006493.1 | - | 1, 2 | 243 | |||
AK016674.1 | - | 1, 2 | 245 | |||
BC049575.1 | - | 1, 2 | 266 | 6773883 | ||
BY094048.1 | - | 1, 2 | 301 | |||
CA464636.1 | - | 1, 2 | 290 |