Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810004N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025615.2 | NP_079891.2 | 2 | 422 | ||
GenBank mRNA | AK011944.1 | - | 2 | 372 | ||
AK012675.1 | - | 2 | 384 | |||
AK017273.1 | - | 2 | 382 | |||
AK165402.1 | - | 2 | 376 | |||
AK166668.1 | - | 2 | 426 | |||
AK166963.1 | - | 2 | 361 | |||
BC023675.1 | - | 2 | 521 | 5354472 | ||
BY030694.1 | - | 2 | 424 |