Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2810004N23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025615.2 | NP_079891.2 | 2 | 157 | ||
GenBank mRNA | AK011944.1 | - | 2 | 107 | ||
AK012675.1 | - | 2 | 119 | |||
AK013989.1 | - | 2 | 145 | |||
AK017273.1 | - | 2 | 117 | |||
AK143919.1 | - | 123 | ||||
AK165402.1 | - | 2 | 111 | |||
AK166668.1 | - | 2 | 161 | |||
AK166963.1 | - | 2 | 96 | |||
BC023675.1 | - | 2 | 256 | 5354472 | ||
BC087916.1 | - | 2 | 140 | |||
BY030694.1 | - | 2 | 157 |