Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipsnap3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025623.2 | NP_079899.1 | 2 | 215 | ||
GenBank mRNA | AF353623.1 | - | 2 | 204 | ||
AK004458.1 | - | 2 | 214 | |||
AK008786.1 | - | 2 | 211 | |||
AK012480.1 | - | 2 | 215 | |||
BC019505.1 | - | 2 | 200 | 4164310 |