Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps19bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175109.3 | NP_780318.1 | 4 | 789 | ||
GenBank mRNA | AB233218.1 | - | 4 | 782 | ||
AK075974.1 | - | 4 | 770 | |||
AV119568.1 | - | 1 | 227 | |||
BC038691.1 | - | 4 | 775 | 1495043 | ||
BC050115.1 | - | 4 | 778 | 30017673 | ||
BC065381.1 | - | 3 | 719 |