Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028260.2 | NP_082536.1 | 5 | 515 | ||
GenBank mRNA | AK005356.1 | - | 5 | 516 | ||
AK009570.1 | - | 5 | 473 | |||
AK010075.1 | - | 5 | 473 | |||
AK011278.1 | - | 5 | 516 | |||
AK011382.1 | - | 5 | 516 | |||
AK011420.1 | - | 5 | 477 | |||
AK011466.1 | - | 5 | 488 | |||
AK011618.1 | - | 5 | 477 | |||
AK011872.1 | - | 5 | 479 | |||
AK012181.1 | - | 468 | ||||
AK015978.1 | - | 5 | 486 | |||
AK078300.1 | - | 5 | 502 | |||
AK135670.1 | - | 5 | 475 | |||
BC008259.1 | - | 5 | 474 | 2649406 | ||
BC081433.1 | - | 4 | 358 | 6814816 |