Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Immp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028260.2 | NP_082536.1 | 6 | 614 | ||
GenBank mRNA | AK005356.1 | - | 6 | 615 | ||
AK009570.1 | - | 6 | 572 | |||
AK010075.1 | - | 6 | 572 | |||
AK011278.1 | - | 6 | 615 | |||
AK011382.1 | - | 6 | 615 | |||
AK011420.1 | - | 6 | 576 | |||
AK011466.1 | - | 6 | 587 | |||
AK011618.1 | - | 6 | 576 | |||
AK011872.1 | - | 6 | 578 | |||
AK012181.1 | - | 567 | ||||
AK015978.1 | - | 6 | 585 | |||
AK078300.1 | - | 6 | 601 | |||
AK135670.1 | - | 6 | 574 | |||
AK168202.1 | - | 6 | 563 | |||
BC008259.1 | - | 6 | 573 | 2649406 | ||
BC081433.1 | - | 5 | 457 | 6814816 |