Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025641.3 | NP_079917.1 | 4 | 463 | ||
GenBank mRNA | AK003266.1 | - | 4 | 428 | ||
AK003539.1 | - | 4 | 406 | |||
AK008953.1 | - | 205 | ||||
AK011734.1 | - | 825 | ||||
AK012552.1 | - | 4 | 441 | |||
AK018712.1 | - | 4 | 441 | |||
AK019085.1 | - | 4 | 320 | |||
AK028137.1 | - | 2 | 205 | |||
AK054005.1 | - | 4 | 429 | |||
AK088185.1 | - | 4 | 428 | |||
BC011388.1 | - | 4 | 407 | 3597862 | ||
BY702808.1 | - | 4 | 436 |