Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025641.3 | NP_079917.1 | 3 | 272 | ||
GenBank mRNA | AK003266.1 | - | 3 | 237 | ||
AK003539.1 | - | 3 | 215 | |||
AK011734.1 | - | 634 | ||||
AK012552.1 | - | 3 | 250 | |||
AK018712.1 | - | 3 | 250 | |||
AK028137.1 | - | 1 | 14 | |||
AK054005.1 | - | 3 | 238 | |||
AK088185.1 | - | 3 | 237 | |||
BY138998.1 | - | 3 | 272 | |||
BY702808.1 | - | 3 | 245 |