Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081090.1 | NP_001074559.1 | 4 | 1175 | ||
GenBank mRNA | AK132230.1 | - | 3 | 1097 | ||
BC010672.1 | - | 4 | 1146 | |||
BC021330.1 | - | 4 | 1194 | |||
BC038028.1 | - | 4 | 517 | |||
BC055464.1 | - | 3 | 1080 | |||
BC096029.1 | - | 4 | 1172 | |||
BC150893.1 | - | 4 | 1141 | 9087804 | ||
BC157979.1 | - | 4 | 1141 | 9007299 |