Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081090.1 | NP_001074559.1 | 14 | 2592 | ||
GenBank mRNA | AK132230.1 | - | 13 | 2514 | ||
BC032932.1 | - | 4 | 500 | |||
BC150893.1 | - | 14 | 2558 | 9087804 | ||
BC157979.1 | - | 14 | 2558 | 9007299 |