Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023229.2 | NP_075718.2 | 6, 7 | 1286 | ||
GenBank mRNA | AF168682.1 | - | 5, 6 | 1181 | ||
AK002552.1 | - | 6, 7 | 1235 | |||
AK003940.1 | - | 3, 4 | 529 | |||
AK038596.1 | - | 6, 7 | 1238 | |||
AK052667.1 | - | 6, 7 | 1231 | |||
AK139374.1 | - | 5, 6 | 2006 | |||
AK151859.1 | - | 6, 7 | 1238 | |||
AK152369.1 | - | 6, 7 | 1287 | |||
AK159751.1 | - | 6, 7 | 1238 | |||
AK164317.1 | - | 1244 | ||||
AK166508.1 | - | 1831 | ||||
AK167346.1 | - | 5, 6 | 1998 | |||
AK177984.1 | - | 2, 3 | 305 | |||
AK215572.1 | - | 1 | 26 | |||
BC013547.1 | - | 6, 7 | 1203 | 4218374 |