Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023230.2 | NP_075719.1 | 3 | 255 | ||
GenBank mRNA | AF303829.1 | - | 2 | 181 | ||
AK002551.1 | - | 3 | 218 | |||
AK012021.1 | - | 3 | 234 | |||
AK158230.1 | - | 3 | 255 | |||
AK170413.1 | - | 3 | 226 | |||
BC003449.1 | - | 3 | 242 | 2654197 | ||
BC092253.1 | - | 3 | 221 | 5055396 |