Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2v1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023230.2 | NP_075719.1 | 2, 3 | 219 | ||
GenBank mRNA | AK002551.1 | - | 2, 3 | 182 | ||
AK012021.1 | - | 2, 3 | 198 | |||
AK158230.1 | - | 2, 3 | 219 | |||
AK170413.1 | - | 2, 3 | 190 | |||
BC003449.1 | - | 2, 3 | 206 | 2654197 | ||
BC092253.1 | - | 2, 3 | 185 | 5055396 |