Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025648.3 | NP_079924.2 | 5 | 602 | ||
GenBank mRNA | AK002577.1 | - | 5 | 579 | ||
AK016506.1 | - | 5 | 598 | |||
AK031697.1 | - | 5 | 580 | |||
AK044637.1 | - | 5 | 580 | |||
AK084031.1 | - | 5 | 564 | |||
AK154629.1 | - | 5 | 600 | |||
AK171872.1 | - | 5 | 483 | |||
AK213018.1 | - | 1 | 66 | |||
BC006862.1 | - | 5 | 573 | 3595108 | ||
BC013533.1 | - | 5 | 571 | 4238911 | ||
CT010350.1 | - | 5 | 553 |