Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025648.3 | NP_079924.2 | 6 | 748 | ||
GenBank mRNA | AK002577.1 | - | 6 | 725 | ||
AK016506.1 | - | 6 | 744 | |||
AK031697.1 | - | 6 | 726 | |||
AK044637.1 | - | 6 | 726 | |||
AK084031.1 | - | 6 | 710 | |||
AK154629.1 | - | 6 | 746 | |||
AK171872.1 | - | 6 | 628 | |||
BC006862.1 | - | 6 | 719 | 3595108 | ||
BC013533.1 | - | 6 | 717 | 4238911 | ||
CT010350.1 | - | 6 | 699 |