Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mad2l1bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025649.3 | NP_079925.2 | 3 | 656 | ||
GenBank mRNA | AK002377.1 | - | 3 | 642 | ||
AK004224.1 | - | 3 | 610 | |||
AK007974.1 | - | 3 | 608 | |||
AK011037.1 | - | 614 | ||||
AK156569.1 | - | 3 | 659 | |||
AK220322.1 | - | 3 | 625 | |||
BC005768.1 | - | 3 | 614 | 2649017 |