Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023232.3 | NP_075721.3 | 3 | 342 | ||
GenBank mRNA | AF203914.1 | - | 3 | 308 | ||
AK002887.1 | - | 3 | 312 | |||
AK012760.1 | - | 3 | 340 | |||
AK077198.1 | - | 3 | 323 | |||
AK082903.1 | - | 3 | 312 | |||
AK133194.1 | - | 322 | ||||
AK169573.1 | - | 3 | 312 | |||
BC024780.1 | - | 3 | 314 | 5365266 | ||
BC054109.1 | - | 3 | 336 | 30013833 |