Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025652.3 | NP_079928.2 | 7 | 812 | ||
GenBank mRNA | AF391799.1 | - | 7 | 796 | ||
AK008247.2 | - | 7 | 780 | |||
AK011845.1 | - | 6 | 1301 | |||
AK012797.1 | - | 7 | 759 | |||
AK017234.1 | - | 7 | 706 | |||
AK143627.1 | - | 7 | 764 | |||
BC032292.1 | - | 6 | 467 | 5374268 | ||
BC057966.1 | - | 6 | 1024 | |||
BC064891.1 | - | 5 | 483 |